Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UHRF1BP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006947.1 | NP_001006948.1 | 12 | 1654 | ||
NM_015054.1 | NP_055869.1 | 12 | 1654 | |||
XM_005268738.2 | XP_005268795.1 | 13 | 1760 | |||
XM_005268739.3 | XP_005268796.1 | 11 | 1431 | |||
XM_011538054.2 | XP_011536356.1 | 2 | 129 | |||
XM_017019047.1 | XP_016874536.1 | 12 | 1614 | |||
XM_017019048.1 | XP_016874537.1 | 12 | 1577 | |||
XM_017019049.1 | XP_016874538.1 | 12 | 1614 | |||
XM_017019050.1 | XP_016874539.1 | 12 | 1559 | |||
XM_017019051.1 | XP_016874540.1 | 13 | 1705 | |||
GenBank mRNA | AB014601.2 | - | 8 | 1095 | ||
AK026472.1 | - | 3 | 251 | |||
AK125813.1 | - | 12 | 1614 | |||
AL136821.1 | - | 5 | 459 | |||
AL833850.1 | - | 2 | 108 | |||
BC014891.1 | - | 12 | 1637 | |||
BC026286.1 | - | 5 | 421 | |||
BC127017.1 | - | 12 | 1472 | 40124660 | ||
BX647380.1 | - | 6 | 625 |