Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129727.2 | NP_001123199.1 | 23 | 4343 | ||
NM_001129728.1 | NP_001123200.1 | 22 | 4303 | |||
NM_001129729.2 | NP_001123201.1 | 22 | 4276 | |||
NM_001129731.2 | NP_001123203.1 | 20 | 4282 | |||
XM_011522985.2 | XP_011521287.1 | 22 | 4273 | |||
XM_011522986.2 | XP_011521288.1 | 22 | 4405 | |||
XM_011522987.2 | XP_011521289.1 | 23 | 4349 | |||
XM_011522988.2 | XP_011521290.1 | 22 | 4222 | |||
GenBank mRNA | AB197663.1 | - | 21 | 6539 | ||
AB197664.1 | - | 20 | 6706 | |||
AK024475.1 | - | 22 | 4238 | |||
AL117435.1 | - | 17 | 3175 | |||
BC001520.2 | - | 7 | 1188 | |||
BC054486.1 | - | 20 | 4217 | 4820741 | ||
BC063501.1 | - | 22 | 4214 | 5527705 | ||
BC082974.1 | - | 23 | 4311 | 6291175 |