Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM63C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020431.3 | NP_065164.2 | 6, 7 | 571 | ||
GenBank mRNA | AK125159.1 | - | 6, 7 | 503 | ||
AL537279.3 | - | 7, 8 | 870 | |||
BC136613.1 | - | 6, 7 | 453 | 9020603 | ||
BC136614.1 | - | 6, 7 | 450 | 9020604 |