Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020715.2 | NP_065766.1 | 29 | 4967 | ||
XM_011537009.1 | XP_011535311.1 | 29 | 5143 | |||
XM_011537015.2 | XP_011535317.1 | 22 | 5198 | |||
XM_017021499.1 | XP_016876988.1 | 28 | 5124 | |||
XM_017021500.1 | XP_016876989.1 | 29 | 5059 | |||
XM_017021501.1 | XP_016876990.1 | 28 | 5040 | |||
XM_017021502.1 | XP_016876991.1 | 22 | 4920 | |||
XM_017021503.1 | XP_016876992.1 | 22 | 4280 | |||
XM_017021504.1 | XP_016876993.1 | 24 | 4422 | |||
GenBank mRNA | AB033026.1 | - | 29 | 4943 | ||
AK123262.1 | - | 2 | 1431 | |||
AK124787.1 | - | 6 | 2024 | |||
BC014159.2 | - | 6 | 1426 | |||
BC037820.1 | - | 1 | 873 | |||
BC146787.1 | - | 29 | 4943 | 9007246 | ||
BX648137.1 | - | 6 | 4299 | |||
BX648464.1 | - | 20 | 4079 |