Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkdcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134117.2 | NP_598878.2 | 7 | 1783 | ||
GenBank mRNA | AB381893.1 | - | 7 | 1775 | ||
AB436780.1 | - | 7 | 1764 | |||
AK133913.1 | - | 3 | 460 | |||
BC022157.1 | - | 6 | 930 | |||
BC086639.1 | - | 8 | 1621 | 6828629 |