Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLSTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014718.3 | NP_055533.2 | 5 | 921 | ||
XM_006719163.3 | XP_006719226.1 | 4 | 2247 | |||
GenBank mRNA | AB018269.1 | - | 4 | 1217 | ||
AJ277460.1 | - | 5 | 918 | |||
AK299177.1 | - | 5 | 895 | |||
AK316059.1 | - | 5 | 921 | |||
AY753302.1 | - | 4 | 679 | |||
BC104767.1 | - | 5 | 725 | 8143770 | ||
BC111491.1 | - | 5 | 909 | 5213830 | ||
BC112283.1 | - | 5 | 737 | 8327751 |