Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014729.2 | NP_055544.1 | 9 | 3235 | ||
XM_017014085.1 | XP_016869574.1 | 8 | 3254 | |||
GenBank mRNA | AB018351.1 | - | 9 | 3237 | ||
CA439630.1 | - | 1 | 540 |