siRNA Details
Gene Symbol: Ggt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002820.1 | NP_001002820.1 | 4 | 1190 | ||
XM_008767848.1 | XP_008766070.1 | 3 | 1379 |

Gene Symbol: Ggt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002820.1 | NP_001002820.1 | 4 | 1190 | ||
XM_008767848.1 | XP_008766070.1 | 3 | 1379 |