Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eml5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003402.2 | NP_001003402.2 | 5 | 949 | ||
XM_003750213.3 | XP_003750261.1 | 5 | 996 | |||
XM_008764827.1 | XP_008763049.1 | 5 | 996 | |||
XM_008764828.1 | XP_008763050.1 | 5 | 997 | |||
GenBank mRNA | AY445136.1 | - | 5 | 957 |