Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003911.2 | NP_001003911.2 | 8 | 1429 | ||
GenBank mRNA | AY551090.1 | - | 8 | 1162 | ||
BC030442.1 | - | 3 | 290 | |||
BC058991.1 | - | 5 | 407 | |||
BC076575.1 | - | 8 | 1283 | |||
BC141173.1 | - | 8 | 1205 | 9055785 | ||
BC145491.1 | - | 8 | 1205 | 9054054 |