Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003912.1 | NP_001003912.1 | 12 | 1035 | ||
GenBank mRNA | AK157225.1 | - | 11 | 2096 | ||
AK165441.1 | - | 11 | 2075 | |||
BC072663.1 | - | 12 | 1685 | |||
BC079565.1 | - | 12 | 1035 | 30536257 | ||
BC079915.1 | - | 12 | 1310 |