Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spin2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005370.1 | NP_001005370.1 | 2 | 315 | ||
GenBank mRNA | BC068162.1 | - | 2 | 315 | 30358613 |