Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270476.1 | NP_001257405.1 | 16 | 2585 | ||
NM_006245.3 | NP_006236.1 | 16 | 2568 | |||
NM_180976.2 | NP_851307.1 | 16 | 2472 | |||
NM_180977.2 | NP_851308.1 | 14 | 2250 | |||
GenBank mRNA | AB209861.1 | - | 13 | 2093 | ||
AK122701.1 | - | 16 | 2462 | |||
BC001095.2 | - | 16 | 2452 | 3507280 | ||
BC001175.1 | - | 16 | 2379 | 3355395 | ||
BC010692.1 | - | 16 | 2462 | 3853892 | ||
D78360.1 | - | 16 | 2370 | |||
L76702.1 | - | 16 | 2576 |