Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RALGAPA1 SIRNA Assays ›
Gene Symbol:
RALGAPA1
Gene Name:
Ral GTPase activating protein catalytic alpha subunit 1
Gene Aliases:
GARNL1, GRIPE, RalGAPalpha1, TULIP1, p240
Chromosome Location:
Chr.14: 35538352 - 35809304 on Build GRCh38
UniGene ID:
Hs.113150
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
222712
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RALGAPA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001283043.1
NP_001269972.1
5, 6
754
NM_001283044.1
NP_001269973.1
5, 6
754
NM_014990.1
NP_055805.1
5, 6
754
NM_194301.2
NP_919277.2
5, 6
754
XM_005267491.3
XP_005267548.1
5, 6
832
XM_005267492.3
XP_005267549.1
5, 6
832
XM_005267493.3
XP_005267550.1
5, 6
832
XM_006720098.2
XP_006720161.1
5, 6
832
XM_006720099.2
XP_006720162.1
5, 6
832
XM_006720100.3
XP_006720163.1
5, 6
832
XM_006720101.2
XP_006720164.1
5, 6
832
XM_006720102.3
XP_006720165.1
5, 6
538
XM_006720103.2
XP_006720166.1
5, 6
832
XM_006720104.3
XP_006720167.1
5, 6
832
XM_011536615.2
XP_011534917.1
5, 6
832
XM_011536616.2
XP_011534918.1
5, 6
832
XM_017021141.1
XP_016876630.1
5, 6
832
XM_017021142.1
XP_016876631.1
5, 6
832
XM_017021143.1
XP_016876632.1
5, 6
832
XM_017021144.1
XP_016876633.1
5, 6
832
XM_017021145.1
XP_016876634.1
5, 6
832
GenBank mRNA
AB511280.1
-
5, 6
363
AY596970.1
-
5, 6
754
AY596971.1
-
5, 6
754
BC150596.1
-
5, 6
462
9056966
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
regulation of small GTPase mediated signal transduction
activation of GTPase activity
GTPase activator activity
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-zvggx:80/10.239.163.62:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0