Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btnl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212489.1 | NP_997654.1 | 1661 | |||
XM_008772738.1 | XP_008770960.1 | 8 | 2053 |
Gene Symbol: Btn3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166344.1 | NP_001159816.1 | 1661 |
Gene Symbol: LOC100911938 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772767.1 | XP_008770989.1 | 1661 |