Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQSEC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134382.2 | NP_001127854.1 | 14 | 5841 | ||
NM_014869.6 | NP_055684.3 | 14 | 3939 | |||
XM_011534304.2 | XP_011532606.1 | 16 | 6078 | |||
XM_011534305.2 | XP_011532607.1 | 14 | 5827 | |||
XM_011534306.2 | XP_011532608.1 | 14 | 6233 | |||
XM_011534307.2 | XP_011532609.1 | 15 | 6179 | |||
XM_011534308.2 | XP_011532610.1 | 14 | 6022 | |||
XM_011534310.2 | XP_011532612.1 | 15 | 6028 | |||
XM_011534311.2 | XP_011532613.1 | 14 | 6190 | |||
XM_011534312.2 | XP_011532614.1 | 14 | 5818 | |||
XM_011534313.2 | XP_011532615.1 | 14 | 6037 | |||
XM_011534314.2 | XP_011532616.1 | 14 | 5773 | |||
XM_011534315.2 | XP_011532617.1 | 13 | 5726 | |||
XM_017007581.1 | XP_016863070.1 | 15 | 6048 | |||
XM_017007582.1 | XP_016863071.1 | 15 | 6033 | |||
GenBank mRNA | AB018306.1 | - | 13 | 3434 | ||
AF055004.1 | - | 1 | 1585 | |||
AY653734.1 | - | 14 | 3699 |