Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010732.4 | NP_034862.1 | 2 | 1411 | ||
GenBank mRNA | AK036271.1 | - | 2 | 1413 | ||
AK039216.1 | - | 2 | 1412 | |||
AY362823.1 | - | 1 | 1000 | |||
BC056458.1 | - | 2 | 1153 | 5685881 | ||
D49375.1 | - | 1 | 119 |