Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025885.3 | NP_080161.1 | 4 | 672 | ||
GenBank mRNA | AK005399.1 | - | 4 | 640 | ||
AK010552.1 | - | 4 | 650 | |||
AK160579.1 | - | 4 | 638 | |||
AK168787.1 | - | 4 | 640 | |||
BB611514.1 | - | 4 | 672 | |||
BC025475.1 | - | 3 | 246 | |||
BC044207.1 | - | 4 | 534 | |||
BC044302.1 | - | 4 | 534 | |||
BC082559.1 | - | 4 | 637 | 30606373 |