Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207633.2 | NP_997516.1 | 6 | 990 | ||
GenBank mRNA | AK020780.1 | - | 6 | 565 | ||
AK030473.1 | - | 6 | 992 | |||
AK045453.1 | - | 6 | 702 | |||
AK053562.1 | - | 6 | 990 | |||
AK079683.1 | - | 6 | 602 | |||
AK143152.1 | - | 6 | 560 | |||
AK160251.1 | - | 7 | 619 | |||
BC048712.1 | - | 6 | 625 | 6328657 |