Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145540.3 | NP_663515.2 | 30 | 3760 | ||
NM_178876.3 | NP_849207.2 | 30 | 3760 | |||
GenBank mRNA | AK033296.1 | - | 30 | 3426 | ||
AK043206.1 | - | 18 | 1699 | |||
AK146699.1 | - | 30 | 3654 | |||
BC003209.1 | - | 16 | 1544 | |||
BC055344.1 | - | 30 | 3295 | 6827687 | ||
BI649325.1 | - | 5 | 375 |