Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007259.2 | NP_001007260.1 | 3 | 298 | ||
NM_001007261.2 | NP_001007262.1 | 4 | 392 | |||
NM_001286752.1 | NP_001273681.1 | 4 | 781 | |||
NM_017438.4 | NP_059134.1 | 3 | 638 | |||
NR_040087.2 | 3 | 638 | ||||
XM_011529636.1 | XP_011527938.1 | 3 | 687 | |||
XM_011529637.1 | XP_011527939.1 | 3 | 752 | |||
XM_011529638.2 | XP_011527940.1 | 3 | 660 | |||
XM_011529639.1 | XP_011527941.1 | 3 | 438 | |||
XM_011529640.2 | XP_011527942.1 | 3 | 703 | |||
XM_011529642.1 | XP_011527944.1 | 4 | 781 | |||
XM_011529643.1 | XP_011527945.1 | 4 | 732 | |||
XM_011529644.1 | XP_011527946.1 | 4 | 293 | |||
XM_017028403.1 | XP_016883892.1 | 3 | 229 | |||
XM_017028404.1 | XP_016883893.1 | 4 | 337 | |||
XM_017028405.1 | XP_016883894.1 | 3 | 638 | |||
GenBank mRNA | AB004848.1 | - | 4 | 345 | ||
AB004852.1 | - | 2 | 169 | |||
AB004853.1 | - | 4 | 341 | |||
AF391112.1 | - | 3 | 205 | |||
AK001660.1 | - | 3 | 205 | |||
AK095215.1 | - | 2 | 1505 | |||
AK300009.1 | - | 3 | 338 | |||
BC002898.2 | - | 3 | 237 | 3940849 | ||
BC036556.2 | - | 4 | 455 | 5266627 | ||
BF195582.1 | - | 3 | 298 | |||
DC368543.1 | - | 4 | 484 |