Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080430.3 | NP_001073899.2 | 7 | 1925 | ||
NM_001146188.2 | NP_001139660.1 | 8 | 1927 | |||
XM_005255892.3 | XP_005255949.1 | 7 | 1948 | |||
XM_011523002.2 | XP_011521304.1 | 7 | 2686 | |||
XM_017023142.1 | XP_016878631.1 | 7 | 2001 | |||
GenBank mRNA | AK025084.1 | - | 1 | 645 | ||
AK027006.1 | - | 6 | 1660 | |||
AK095095.1 | - | 2 | 2220 | |||
AK299202.1 | - | 8 | 1927 | |||
U80736.1 | - | 3 | 1019 |