Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102664.1 | NP_001096134.1 | 8 | 2365 | ||
NM_014964.4 | NP_055779.2 | 11 | 3424 | |||
NM_148921.3 | NP_683723.2 | 10 | 3253 | |||
GenBank mRNA | AB028988.1 | - | 11 | 3389 |