Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OC90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080399.2 | NP_001073868.2 | 8 | 698 | ||
GenBank mRNA | AK297911.1 | - | 9 | 698 |