Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFHX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033400.2 | NP_207646.2 | 10 | 8043 | ||
XM_005268135.3 | XP_005268192.1 | 6 | 5206 | |||
XM_011537245.2 | XP_011535547.1 | 10 | 7963 | |||
XM_011537246.2 | XP_011535548.1 | 10 | 10399 | |||
XM_011537247.2 | XP_011535549.1 | 10 | 8978 | |||
XM_011537252.2 | XP_011535554.1 | 6 | 5181 | |||
XM_017021714.1 | XP_016877203.1 | 10 | 7958 | |||
XM_017021715.1 | XP_016877204.1 | 10 | 7885 | |||
XM_017021716.1 | XP_016877205.1 | 9 | 7723 | |||
XM_017021718.1 | XP_016877207.1 | 7 | 4982 | |||
GenBank mRNA | AB051549.1 | - | 2 | 4351 | ||
BC144336.1 | - | 1 | 4809 | 9052856 | ||
BC150606.1 | - | 2 | 4548 | 9056976 |