Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BICC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080512.2 | NP_001073981.1 | 12 | 1551 | ||
XM_005270169.4 | XP_005270226.1 | 11 | 4549 | |||
XM_011540185.2 | XP_011538487.1 | 13 | 1797 | |||
XM_011540188.2 | XP_011538490.2 | 12 | 1682 | |||
XM_011540189.2 | XP_011538491.1 | 11 | 1448 | |||
XM_011540190.2 | XP_011538492.1 | 11 | 1405 | |||
XM_011540191.2 | XP_011538493.1 | 9 | 1267 | |||
XM_017016677.1 | XP_016872166.1 | 13 | 8185 | |||
XM_017016678.1 | XP_016872167.1 | 14 | 1702 | |||
GenBank mRNA | AK307780.1 | - | 6 | 835 | ||
AY726586.1 | - | 12 | 1496 |