Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSHZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193421.1 | NP_001180350.1 | 3 | 5806 | ||
NM_173485.5 | NP_775756.3 | 3 | 6339 | |||
XM_017027641.1 | XP_016883130.1 | 3 | 6310 | |||
GenBank mRNA | AF230201.1 | - | 1 | 528 | ||
AY926481.1 | - | 1 | 531 |