Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015205.2 | NP_056020.2 | 30 | 6771 | ||
NM_032189.3 | NP_115565.3 | 29 | 6744 | |||
XM_005268299.4 | XP_005268356.1 | 29 | 7042 | |||
XM_005268300.4 | XP_005268357.1 | 29 | 6978 | |||
XM_005268305.4 | XP_005268362.1 | 30 | 7128 | |||
XM_005268306.4 | XP_005268363.1 | 30 | 7125 | |||
GenBank mRNA | AB028944.2 | - | 29 | 6648 | ||
AK025511.1 | - | 1 | 483 | |||
AL161996.1 | - | 13 | 4814 |