Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELOVL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104558.1 | NP_001098028.1 | 5, 6 | 538 | ||
NM_001297617.1 | NP_001284546.1 | 8, 9 | 936 | |||
NM_001297618.1 | NP_001284547.1 | 6, 7 | 543 | |||
NM_024930.2 | NP_079206.2 | 6, 7 | 589 | |||
XM_005248606.4 | XP_005248663.1 | 5, 6 | 1153 | |||
XM_005248607.3 | XP_005248664.1 | 6, 7 | 866 | |||
XM_006714695.3 | XP_006714758.1 | 7, 8 | 645 | |||
XM_011543651.2 | XP_011541953.1 | 7, 8 | 835 | |||
XM_017009885.1 | XP_016865374.1 | 6, 7 | 526 | |||
XM_017009886.1 | XP_016865375.1 | 8, 9 | 765 | |||
XM_017009887.1 | XP_016865376.1 | 8, 9 | 956 | |||
GenBank mRNA | AB181393.1 | - | 6, 7 | 530 | ||
AK027216.1 | - | 4, 5 | 358 | |||
AL137506.1 | - | 1, 2 | 20 | |||
BC094792.1 | - | 8, 9 | 877 | |||
BC130310.1 | - | 5, 6 | 486 | 40146341 | ||
BC130312.1 | - | 5, 6 | 486 | 40146343 | ||
DA247166.1 | - | 5, 6 | 538 |