Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRTG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173814.5 | NP_776175.2 | 20 | 3717 | ||
XM_017022081.1 | XP_016877570.1 | 20 | 3804 | |||
GenBank mRNA | AK098622.1 | - | 11 | 1725 | ||
AY630258.1 | - | 20 | 3577 |