Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GNTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009905.2 | NP_001009905.1 | 10 | 893 | ||
NM_001320742.1 | NP_001307671.1 | 11 | 1011 | |||
NM_001320743.1 | NP_001307672.1 | 6 | 1488 | |||
NR_135465.1 | 10 | 893 | ||||
NR_135466.1 | 6 | 1479 | ||||
XM_006722272.3 | XP_006722335.1 | 10 | 927 | |||
XM_011523532.2 | XP_011521834.1 | 9 | 861 | |||
XM_011523535.1 | XP_011521837.1 | 7 | 1830 | |||
XM_017024205.1 | XP_016879694.1 | 9 | 831 | |||
XM_017024206.1 | XP_016879695.1 | 7 | 614 | |||
GenBank mRNA | AK054757.1 | - | 6 | 1479 | ||
AY304503.1 | - | 10 | 891 | |||
AY634364.1 | - | 10 | 893 | |||
BC019623.1 | - | 8 | 687 |