Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL21B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015433.2 | NP_056248.2 | 3 | 856 | ||
NM_206914.1 | NP_996797.1 | 4 | 995 | |||
GenBank mRNA | AF455816.1 | - | 3 | 856 | ||
AK024983.1 | - | 4 | 883 | |||
AK292276.1 | - | 3 | 800 | |||
AL050100.1 | - | 4 | 917 | |||
BC016395.1 | - | 3 | 856 | 3882414 | ||
BC099841.1 | - | 3 | 831 | 40010149 | ||
BC103501.1 | - | 3 | 831 | 40010152 |