Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271.3 | NP_001262.3 | 39 | 7436 | ||
GenBank mRNA | AL831836.1 | - | 1 | 1676 | ||
BX537626.1 | - | 1 | 253 |