Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000254.2 | NP_000245.2 | 33 | 7601 | ||
NM_001291939.1 | NP_001278868.1 | 32 | 7448 | |||
NM_001291940.1 | NP_001278869.1 | 32 | 7488 | |||
XM_005273141.4 | XP_005273198.1 | 33 | 7570 | |||
XM_006711770.2 | XP_006711833.1 | 27 | 6549 | |||
XM_011544194.2 | XP_011542496.1 | 32 | 8363 | |||
XM_017001329.1 | XP_016856818.1 | 31 | 8210 | |||
XM_017001330.1 | XP_016856819.1 | 30 | 8174 | |||
GenBank mRNA | AK125206.1 | - | 1 | 105 | ||
BC015894.1 | - | 1 | 57 |