Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B9D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243473.2 | NP_001230402.1 | 6 | 940 | ||
NM_001321214.1 | NP_001308143.1 | 7 | 794 | |||
NM_001321215.1 | NP_001308144.1 | 6 | 1033 | |||
NM_015681.4 | NP_056496.1 | 7 | 705 | |||
XM_005256607.2 | XP_005256664.1 | 7 | 711 | |||
GenBank mRNA | AB030506.1 | - | 7 | 694 | ||
AK293815.1 | - | 6 | 1006 |