Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015715.4 | NP_056530.2 | 2, 3 | 908 | ||
XM_011530204.1 | XP_011528506.1 | 3 | 1130 | |||
XM_011530205.1 | XP_011528507.1 | 3 | 1119 | |||
GenBank mRNA | AF220490.1 | - | 2, 3 | 908 | ||
BC025316.1 | - | 2, 3 | 784 | 5458919 |