Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015982.3 | NP_057066.2 | 3 | 413 | ||
XM_017024713.1 | XP_016880202.1 | 3 | 458 | |||
GenBank mRNA | AB208818.1 | - | 1 | 2551 | ||
AF096834.1 | - | 3 | 411 | |||
AK092747.1 | - | 3 | 187 | |||
AK315035.1 | - | 3 | 384 | |||
BC033800.1 | - | 3 | 376 | 5171696 | ||
BC047760.1 | - | 3 | 377 | 5742121 |