Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUSC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172367.2 | NP_758955.2 | 3 | 2940 | ||
GenBank mRNA | AL831929.1 | - | 4 | 2336 | ||
BC142615.1 | - | 3 | 2924 | 40147346 |