Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPNMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005340.1 | NP_001005340.1 | 5 | 815 | ||
NM_002510.2 | NP_002501.1 | 5 | 815 | |||
XM_005249578.2 | XP_005249635.1 | 5 | 1306 | |||
XM_017011676.1 | XP_016867165.1 | 5 | 1306 | |||
XM_017011677.1 | XP_016867166.1 | 5 | 1306 | |||
XM_017011678.1 | XP_016867167.1 | 5 | 1306 | |||
GenBank mRNA | AF322909.1 | - | 5 | 713 | ||
AK292078.1 | - | 5 | 737 | |||
AK296779.1 | - | 5 | 600 | |||
AK297031.1 | - | 4 | 563 | |||
AK297051.1 | - | 5 | 584 | |||
AK297055.1 | - | 4 | 393 | |||
AK297803.1 | - | 3 | 440 | |||
AK304671.1 | - | 6 | 796 | |||
AK312279.1 | - | 5 | 744 | |||
AY359124.1 | - | 5 | 735 | |||
BC025297.1 | - | 5 | 705 | |||
BC032783.1 | - | 5 | 731 | 5177095 | ||
X76534.1 | - | 5 | 745 |