Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MON2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278469.1 | NP_001265398.1 | 36 | 5548 | ||
NM_001278470.1 | NP_001265399.1 | 34 | 5469 | |||
NM_001278471.1 | NP_001265400.1 | 34 | 5400 | |||
NM_001278472.1 | NP_001265401.1 | 36 | 5565 | |||
NM_015026.2 | NP_055841.2 | 35 | 5487 | |||
XM_017019041.1 | XP_016874530.1 | 36 | 5490 | |||
XM_017019042.1 | XP_016874531.1 | 35 | 5472 | |||
XM_017019043.1 | XP_016874532.1 | 37 | 5564 | |||
XM_017019044.1 | XP_016874533.1 | 37 | 5566 | |||
XM_017019045.1 | XP_016874534.1 | 38 | 5234 | |||
XM_017019046.1 | XP_016874535.1 | 32 | 4659 | |||
GenBank mRNA | AB017814.1 | - | 35 | 5212 | ||
AB028963.3 | - | 36 | 5458 | |||
AK022671.1 | - | 9 | 1664 | |||
AK092646.1 | - | 9 | 2251 | |||
AL833066.2 | - | 36 | 5543 | |||
AL834320.1 | - | 29 | 4445 | |||
BC136621.1 | - | 34 | 5438 | 9020611 | ||
BC144309.1 | - | 34 | 5369 | 9052829 | ||
BC151241.1 | - | 36 | 5481 | 8860514 | ||
BX537415.1 | - | 35 | 5485 |