Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242466.1 | NP_001229395.1 | 9 | 4026 | ||
NM_181504.3 | NP_852556.2 | 10 | 4163 | |||
NM_181523.2 | NP_852664.1 | 16 | 5463 | |||
NM_181524.1 | NP_852665.1 | 10 | 4295 | |||
XM_005248542.3 | XP_005248599.1 | 16 | 5486 | |||
XM_011543493.2 | XP_011541795.1 | 15 | 5116 | |||
XM_017009585.1 | XP_016865074.1 | 16 | 6255 | |||
XM_017009586.1 | XP_016865075.1 | 15 | 5076 | |||
GenBank mRNA | BE888150.1 | - | 1 | 178 | ||
BQ723333.1 | - | 1 | 621 |