Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRTM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015564.2 | NP_056379.1 | 2 | 1621 | ||
GenBank mRNA | AB007876.1 | - | 2 | 1573 | ||
AK291064.1 | - | 2 | 1592 | |||
AK297332.1 | - | 2 | 1167 | |||
AY182026.1 | - | 1 | 1164 | |||
BC126408.1 | - | 2 | 1192 | 8992124 | ||
BC126410.1 | - | 2 | 1192 | 8992126 | ||
BC143926.1 | - | 2 | 1192 | 9052444 |