Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002282.3 | NP_002273.3 | 1 | 76 | ||
GenBank mRNA | AK314904.1 | - | 1 | 76 | ||
BC069546.1 | - | 1 | 63 | 7262357 | ||
BC126935.1 | - | 1 | 35 | 40018026 | ||
BC128063.1 | - | 1 | 35 | 40006347 | ||
X99141.1 | - | 65 |