Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOBEC3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021822.3 | NP_068594.1 | 2 | 433 | ||
XM_017028903.1 | XP_016884392.1 | 2 | 433 | |||
GenBank mRNA | AB266487.1 | - | 2 | 76 | ||
AF182420.1 | - | 2 | 307 | |||
AK310279.1 | - | 2 | 186 | |||
AK315650.1 | - | 2 | 359 | |||
BC024268.1 | - | 2 | 118 | 4877863 | ||
BC061914.1 | - | 2 | 317 | |||
CR456472.1 | - | 2 | 180 | |||
DA457434.1 | - | 2 | 418 | |||
JF262036.1 | - | 2 | 76 | |||
JX915801.1 | - | 2 | 76 | |||
KC329531.1 | - | 2 | 76 |
Gene Symbol: APOBEC3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006666.1 | NP_001006667.1 | 2 | 369 | ||
NM_145298.5 | NP_660341.2 | 2 | 369 | |||
XM_011529994.2 | XP_011528296.1 | 2 | 433 | |||
XM_017028642.1 | XP_016884131.1 | 2 | 130 | |||
XM_017028643.1 | XP_016884132.1 | 3 | 824 | |||
GenBank mRNA | AK294810.1 | - | 2 | 433 | ||
BC038808.1 | - | 2 | 154 | 5227163 | ||
BG280786.1 | - | 153 | ||||
BG758984.1 | - | 2 | 332 | |||
BQ056344.1 | - | 2 | 402 | |||
CR456395.1 | - | 2 | 180 | |||
CX165430.1 | - | 2 | 160 | |||
DA221480.1 | - | 2 | 433 | |||
JF262037.1 | - | 2 | 76 | |||
KC329530.1 | - | 2 | 76 |