Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080461.1 | NP_001073930.1 | 2 | 331 | ||
GenBank mRNA | KM587717.1 | - | 2 | 967 | ||
KM587718.1 | - | 2 | 501 | |||
KM587719.1 | - | 2 | 501 |