Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNMT3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320893.1 | NP_001307822.1 | 16 | 2008 | ||
NM_022552.4 | NP_072046.2 | 21 | 2694 | |||
NM_153759.3 | NP_715640.2 | 17 | 1978 | |||
NM_175629.2 | NP_783328.1 | 21 | 2765 | |||
NR_135490.1 | 22 | 2964 | ||||
XM_005264175.4 | XP_005264232.1 | 21 | 2628 | |||
XM_005264177.4 | XP_005264234.1 | 16 | 1920 | |||
XM_011532662.2 | XP_011530964.1 | 19 | 2630 | |||
XM_011532663.2 | XP_011530965.1 | 22 | 2807 | |||
XM_011532666.2 | XP_011530968.1 | 17 | 1993 | |||
XM_011532667.2 | XP_011530969.1 | 16 | 1810 | |||
XM_017003526.1 | XP_016859015.1 | 21 | 2736 | |||
XM_017003527.1 | XP_016859016.1 | 16 | 1964 | |||
GenBank mRNA | AB208833.1 | - | 22 | 2863 | ||
AF067972.2 | - | 21 | 2656 | |||
AF331856.1 | - | 21 | 2644 | |||
AF480163.1 | - | 17 | 1974 | |||
AK025230.1 | - | 11 | 1018 | |||
BC018214.1 | - | 6 | 547 | |||
BC023612.2 | - | 7 | 746 | |||
BC043617.1 | - | 21 | 2664 | 6150112 |