Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POFUT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015227.4 | NP_056042.1 | 8 | 4161 | ||
NM_133635.4 | NP_598368.2 | 9 | 2207 | |||
NR_004858.1 | 10 | 2413 | ||||
XM_006723981.3 | XP_006724044.1 | 10 | 2280 | |||
XM_011529504.2 | XP_011527806.1 | 11 | 2416 | |||
XM_011529506.2 | XP_011527808.1 | 10 | 2355 | |||
XM_011529507.2 | XP_011527809.1 | 10 | 2251 | |||
XM_011529510.2 | XP_011527812.1 | 11 | 2325 | |||
XM_017028306.1 | XP_016883795.1 | 9 | 2190 | |||
XM_017028309.1 | XP_016883798.1 | 7 | 1613 | |||
GenBank mRNA | AB023175.1 | - | 8 | 4151 | ||
AJ302079.2 | - | 10 | 2402 | |||
AJ302080.2 | - | 8 | 4150 | |||
AL110285.1 | - | 10 | 2399 | |||
AY066015.2 | - | 9 | 2196 | |||
BC011044.1 | - | 6 | 1861 | |||
BC064623.1 | - | 9 | 2189 | 5727996 |