Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318498.1 | NP_001305427.1 | 3 | 196 | ||
NM_001318499.1 | NP_001305428.1 | 3 | 196 | |||
NM_001318500.1 | NP_001305429.1 | 3 | 196 | |||
NM_016055.5 | NP_057139.1 | 3 | 196 | |||
NR_134659.1 | 5 | 449 | ||||
NR_134661.1 | 3 | 196 | ||||
XM_017017919.1 | XP_016873408.1 | 3 | 164 | |||
GenBank mRNA | AF151876.1 | - | 3 | 752 | ||
AF161408.1 | - | 3 | 199 | |||
AK297746.1 | - | 3 | 196 | |||
AK310005.1 | - | 5 | 428 | |||
BC009764.1 | - | 3 | 189 | 4093230 | ||
BM817807.1 | - | 3 | 194 | |||
BM828996.1 | - | 3 | 195 | |||
BP210564.1 | - | 3 | 174 | |||
BU600573.1 | - | 3 | 160 | |||
BU932890.1 | - | 3 | 195 | |||
DA549653.1 | - | 3 | 196 |