Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPSAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003294.3 | NP_003285.2 | 6 | 893 | ||
GenBank mRNA | AF206667.1 | - | 4 | 655 | ||
BC038416.1 | - | 5 | 847 | |||
BC065923.1 | - | 5 | 1052 | 5752632 | ||
BC074975.2 | - | 6 | 874 | 30915294 | ||
FJ931118.1 | - | 4 | 690 | |||
M33491.1 | - | 5 | 837 |
Gene Symbol: TPSB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC029356.1 | - | 6 | 864 | 4695458 | |
M33492.1 | - | 5 | 839 | |||
S55551.1 | - | 5 | 848 |