Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001843.3 | NP_001834.2 | 22 | 3016 | ||
NM_175038.2 | NP_778203.1 | 20 | 2832 | |||
XM_005268651.2 | XP_005268708.1 | 22 | 3379 | |||
XM_006719241.2 | XP_006719304.1 | 22 | 3003 | |||
XM_011537926.2 | XP_011536228.1 | 22 | 2968 | |||
XM_011537927.2 | XP_011536229.1 | 22 | 3025 | |||
XM_017018821.1 | XP_016874310.1 | 22 | 3019 | |||
XM_017018822.1 | XP_016874311.1 | 22 | 2968 | |||
XM_017018823.1 | XP_016874312.1 | 22 | 3379 | |||
XM_017018824.1 | XP_016874313.1 | 22 | 3003 | |||
GenBank mRNA | U07819.1 | - | 21 | 2789 | ||
U07820.1 | - | 20 | 2837 | |||
Z21488.1 | - | 22 | 2910 |
Set Membership: |
G-protein coupled receptor |